siRNA Details
| Gene Symbol: Klf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031642.4 | NP_113830.2 | 2 | 643 | ||
| XM_006254182.1 | XP_006254244.1 | 9 | 1909 | |||
| GenBank mRNA | AF001417.1 | - | 2 | 423 | ||
| AY318957.1 | - | 9 | 1908 | |||
| BC097306.1 | - | 2 | 568 | 7444817 | ||
| FQ225029.1 | - | 1 | 199 | |||
| FQ230654.1 | - | 1 | 293 | |||

Klf6
Genomic Map