siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053322.2 | NP_445774.1 | 38, 39 | 5502 | ||
| XM_006236921.2 | XP_006236983.1 | 36, 37 | 5683 | |||
| GenBank mRNA | Y00826.1 | - | 38, 39 | 5503 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup210| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053322.2 | NP_445774.1 | 38, 39 | 5502 | ||
| XM_006236921.2 | XP_006236983.1 | 36, 37 | 5683 | |||
| GenBank mRNA | Y00826.1 | - | 38, 39 | 5503 | ||