siRNA Details
| Gene Symbol: Map2k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133283.1 | NP_579817.1 | 3 | 420 | ||
| XM_006240987.2 | XP_006241049.1 | 3 | 607 | |||
| GenBank mRNA | BC126084.1 | - | 3 | 518 | 8376547 | |
| D14592.1 | - | 3 | 391 | |||
| L14936.1 | - | 3 | 420 | |||

Map2k2
Genomic Map