siRNA Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301702.1 | NP_001288631.1 | 12 | 1877 | ||
| NM_022585.2 | NP_072107.2 | 13 | 2035 | |||
| XM_006241568.2 | XP_006241630.1 | 13 | 2008 | |||
| XM_006241569.2 | XP_006241631.1 | 12 | 1869 | |||
| XM_008765457.1 | XP_008763679.1 | 12 | 1872 | |||
| GenBank mRNA | BC061550.1 | - | 10 | 1273 | 6918922 | |
| D50734.1 | - | 12 | 1670 | |||
| D89983.1 | - | 13 | 1983 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001102458, NM_018745, AK088671, BC043722, AK049680, and others.

Azin1
Genomic Map