siRNA Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301702.1 | NP_001288631.1 | 10 | 1563 | ||
| NM_022585.2 | NP_072107.2 | 11 | 1721 | |||
| XM_006241568.2 | XP_006241630.1 | 11 | 1694 | |||
| XM_006241569.2 | XP_006241631.1 | 10 | 1555 | |||
| XM_008765457.1 | XP_008763679.1 | 10 | 1558 | |||
| GenBank mRNA | BC061550.1 | - | 8 | 959 | 6918922 | |
| D50734.1 | - | 10 | 1356 | |||
| D89983.1 | - | 11 | 1669 | |||

Azin1
Genomic Map