siRNA Details
| Gene Symbol: Hpgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031644.2 | NP_113832.1 | 6 | 902 | ||
| XM_006236602.2 | XP_006236664.1 | 6 | 873 | |||
| XM_006236603.2 | XP_006236665.1 | 5 | 795 | |||
| GenBank mRNA | AF021882.1 | - | 5 | 430 | ||
| BC087590.1 | - | 6 | 902 | 7307804 | ||
| D82071.1 | - | 6 | 733 | |||

Hpgds
Genomic Map