siRNA Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191884.1 | NP_001178813.1 | 3 | 284 | ||
| XM_006243847.2 | XP_006243909.1 | 3 | 293 | |||
| XM_006243848.2 | XP_006243910.1 | 3 | 219 | |||
| XM_006243849.1 | XP_006243911.1 | 3 | 737 | |||
| XM_006243850.2 | XP_006243912.1 | 3 | 258 | |||
| XM_006243851.1 | XP_006243913.1 | 3 | 322 | |||
| GenBank mRNA | AF051943.1 | - | 1 | 26 | ||
| FQ225386.1 | - | 2 | 214 | |||

Nme6
Genomic Map