siRNA Details
Gene Symbol: Nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191884.1 | NP_001178813.1 | 4 | 352 | ||
XM_006243847.2 | XP_006243909.1 | 4 | 361 | |||
XM_006243848.2 | XP_006243910.1 | 4 | 287 | |||
XM_006243849.1 | XP_006243911.1 | 4 | 805 | |||
XM_006243850.2 | XP_006243912.1 | 4 | 326 | |||
XM_006243851.1 | XP_006243913.1 | 4 | 390 | |||
GenBank mRNA | AF051943.1 | - | 2 | 94 | ||
FQ225386.1 | - | 3 | 442 |
