siRNA Details
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031647.2 | NP_113835.2 | 6 | 681 | ||
XM_006252656.2 | XP_006252718.1 | 6 | 967 | |||
XM_006252657.2 | XP_006252719.1 | 7 | 1027 | |||
XM_006252658.1 | XP_006252720.1 | 6 | 2939 | |||
XM_008771003.1 | XP_008769225.1 | 8 | 1948 | |||
XM_008771004.1 | XP_008769226.1 | 7 | 1888 | |||
XM_008771005.1 | XP_008769227.1 | 9 | 1257 | |||
XM_008771006.1 | XP_008769228.1 | 8 | 1121 | |||
GenBank mRNA | AB032164.1 | - | 6 | 675 | ||
BC081770.1 | - | 6 | 681 | 7132495 |
