siRNA Details
| Gene Symbol: Acan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022190.1 | NP_071526.1 | 12 | 5326 | ||
| GenBank mRNA | J03485.1 | - | 12 | 5326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acan| Gene Symbol: Acan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022190.1 | NP_071526.1 | 12 | 5326 | ||
| GenBank mRNA | J03485.1 | - | 12 | 5326 | ||