siRNA Details
| Gene Symbol: Gabarap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172036.3 | NP_742033.1 | 3 | 274 | ||
| GenBank mRNA | AF161588.1 | - | 3 | 288 | ||
| BC058441.1 | - | 3 | 278 | 6918795 | ||
| FQ213199.1 | - | 3 | 345 | |||
| FQ224284.1 | - | 3 | 240 | |||
| FQ229256.1 | - | 3 | 276 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019749, AF161587, BC024621, AK002879, AK011731, and others.

Gabarap
Genomic Map