siRNA Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031649.2 | NP_113837.1 | 4, 5 | 457 | ||
| XM_006237313.2 | XP_006237375.1 | 3, 4 | 420 | |||
| GenBank mRNA | X79812.1 | - | 4, 5 | 510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrg1| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031649.2 | NP_113837.1 | 4, 5 | 457 | ||
| XM_006237313.2 | XP_006237375.1 | 3, 4 | 420 | |||
| GenBank mRNA | X79812.1 | - | 4, 5 | 510 | ||