siRNA Details
| Gene Symbol: Ppp1r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022676.3 | NP_073167.1 | 5 | 396 | ||
| XM_006242437.2 | XP_006242499.1 | 5 | 408 | |||
| GenBank mRNA | AJ276593.1 | - | 5 | 269 | ||
| AY648296.1 | - | 5 | 396 | |||
| BC078820.1 | - | 5 | 397 | 7127505 | ||
| FQ217772.1 | - | 5 | 365 | |||
| J05592.1 | - | 5 | 268 | |||

Ppp1r1a
Genomic Map