siRNA Details
| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031653.2 | NP_113841.2 | 8 | 2048 | ||
| GenBank mRNA | AF060879.1 | - | 3 | 1150 | ||
| M97161.1 | - | 8 | 2030 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncan| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031653.2 | NP_113841.2 | 8 | 2048 | ||
| GenBank mRNA | AF060879.1 | - | 3 | 1150 | ||
| M97161.1 | - | 8 | 2030 | |||