siRNA Details
| Gene Symbol: Grk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022928.1 | NP_075217.1 | 8, 9 | 924 | ||
| XM_006251359.1 | XP_006251421.1 | 7, 8 | 667 | |||
| XM_006251360.1 | XP_006251422.1 | 8, 9 | 760 | |||
| XM_008770353.1 | XP_008768575.1 | 8, 9 | 760 | |||
| XM_008770354.1 | XP_008768576.1 | 7, 8 | 667 | |||
| XM_008770355.1 | XP_008768577.1 | 8, 9 | 760 | |||
| XM_008770357.1 | XP_008768579.1 | 3, 4 | 315 | |||
| GenBank mRNA | X97568.1 | - | 8, 9 | 924 | ||

Grk4
Genomic Map