siRNA Details
| Gene Symbol: Asl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021577.3 | NP_067588.2 | 9 | 1171 | ||
| XM_006249245.2 | XP_006249307.1 | 8 | 915 | |||
| XM_006249247.2 | XP_006249309.1 | 7 | 857 | |||
| GenBank mRNA | BC078682.1 | - | 7 | 794 | 7109106 | |
| BC097270.1 | - | 9 | 1171 | 7455735 | ||
| D13978.1 | - | 7 | 1519 | |||
| D28501.1 | - | 7 | 702 | |||

Asl
Genomic Map