siRNA Details
| Gene Symbol: P3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021581.1 | NP_067592.1 | 3, 4 | 730 | ||
| GenBank mRNA | X65454.1 | - | 3, 4 | 730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P3h4| Gene Symbol: P3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021581.1 | NP_067592.1 | 3, 4 | 730 | ||
| GenBank mRNA | X65454.1 | - | 3, 4 | 730 | ||