siRNA Details
| Gene Symbol: Ptges | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021583.3 | NP_067594.1 | 2 | 230 | ||
| GenBank mRNA | AB035145.1 | - | 2 | 190 | ||
| AB041998.1 | - | 2 | 219 | |||
| AB048730.1 | - | 2 | 232 | |||
| AF280967.1 | - | 2 | 229 | |||
| BC088101.1 | - | 2 | 188 | |||
| BC105858.1 | - | 2 | 216 | 7456259 | ||

Ptges
Genomic Map