siRNA Details
| Gene Symbol: Milr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021585.1 | NP_067596.1 | 1 | 162 | ||
| XM_008768388.1 | XP_008766610.1 | 1 | 310 | |||
| XM_008768389.1 | XP_008766611.1 | 1 | 310 | |||
| GenBank mRNA | U39546.1 | - | 1 | 162 | ||

Milr1
Genomic Map