siRNA Details
| Gene Symbol: Aipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021590.1 | NP_067601.1 | 2 | 187 | ||
| XM_006246798.2 | XP_006246860.1 | 2 | 219 | |||
| GenBank mRNA | AF180340.1 | - | 2 | 187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aipl1| Gene Symbol: Aipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021590.1 | NP_067601.1 | 2 | 187 | ||
| XM_006246798.2 | XP_006246860.1 | 2 | 219 | |||
| GenBank mRNA | AF180340.1 | - | 2 | 187 | ||