siRNA Details
| Gene Symbol: Ninj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021595.2 | NP_067606.1 | 3 | 415 | ||
| XM_008763255.1 | XP_008761477.1 | 3 | 925 | |||
| XM_008763256.1 | XP_008761478.1 | 4 | 842 | |||
| XM_008763257.1 | XP_008761479.1 | 4 | 601 | |||
| XM_008763258.1 | XP_008761480.1 | 4 | 752 | |||
| XM_008763259.1 | XP_008761481.1 | 3 | 561 | |||
| GenBank mRNA | AB040815.1 | - | 3 | 416 | ||
| AF250322.1 | - | 3 | 416 | |||

Ninj2
Genomic Map