siRNA Details
| Gene Symbol: Ltb4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021656.1 | NP_067688.1 | 1 | 389 | ||
| XM_006252011.2 | XP_006252073.1 | 2 | 2063 | |||
| XM_006252012.1 | XP_006252074.1 | 2 | 425 | |||
More Information
Set Membership: |
|

Ltb4r
Genomic Map