siRNA Details
| Gene Symbol: Ip6k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021660.2 | NP_067692.2 | 5 | 830 | ||
| XM_008766564.1 | XP_008764786.1 | 4 | 796 | |||
| XM_008766565.1 | XP_008764787.1 | 3 | 506 | |||
| XM_008766566.1 | XP_008764788.1 | 5 | 865 | |||
| GenBank mRNA | BC083574.1 | - | 5 | 830 | 7112777 | |

Ip6k2
Genomic Map