siRNA Details
| Gene Symbol: Ip6k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021660.2 | NP_067692.2 | 4 | 548 | ||
| XM_008766564.1 | XP_008764786.1 | 3 | 514 | |||
| XM_008766565.1 | XP_008764787.1 | 2 | 224 | |||
| XM_008766566.1 | XP_008764788.1 | 4 | 583 | |||
| GenBank mRNA | BC083574.1 | - | 4 | 548 | 7112777 | |

Ip6k2
Genomic Map