siRNA Details
| Gene Symbol: Bmp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021670.1 | NP_067702.1 | 2 | 1007 | ||
| GenBank mRNA | AJ132407.1 | - | 2 | 1007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp15| Gene Symbol: Bmp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021670.1 | NP_067702.1 | 2 | 1007 | ||
| GenBank mRNA | AJ132407.1 | - | 2 | 1007 | ||