siRNA Details
| Gene Symbol: Tmem33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034198.2 | NP_001029370.1 | 6, 7 | 694 | ||
| NM_021671.3 | NP_067703.1 | 6, 7 | 694 | |||
| GenBank mRNA | AB006135.1 | - | 6, 7 | 652 | ||
| BF552634.1 | - | 1, 2 | 26 | |||
| CK359233.1 | - | 6, 7 | 575 | |||

Tmem33
Genomic Map