siRNA Details
| Gene Symbol: Tcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021673.1 | NP_067705.1 | 4 | 763 | ||
| XM_006247642.2 | XP_006247704.1 | 5 | 1517 | |||
| GenBank mRNA | BC166495.1 | - | 5 | 825 | 7111123 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcam1| Gene Symbol: Tcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021673.1 | NP_067705.1 | 4 | 763 | ||
| XM_006247642.2 | XP_006247704.1 | 5 | 1517 | |||
| GenBank mRNA | BC166495.1 | - | 5 | 825 | 7111123 | |