siRNA Details
| Gene Symbol: Psgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021677.1 | NP_067709.1 | 3 | 488 | ||
| XM_006228464.1 | XP_006228526.1 | 2 | 357 | |||
| GenBank mRNA | U66292.1 | - | 3 | 488 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psgb1| Gene Symbol: Psgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021677.1 | NP_067709.1 | 3 | 488 | ||
| XM_006228464.1 | XP_006228526.1 | 2 | 357 | |||
| GenBank mRNA | U66292.1 | - | 3 | 488 | ||