siRNA Details
| Gene Symbol: Nxph4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021680.2 | NP_067712.2 | 2 | 864 | ||
| XM_008765418.1 | XP_008763640.1 | 2 | 860 | |||
| GenBank mRNA | AF042714.1 | - | 2 | 935 | ||
| BC081805.1 | - | 2 | 864 | 7108207 | ||

Nxph4
Genomic Map