siRNA Details
| Gene Symbol: Adcy10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021684.1 | NP_067716.1 | 7 | 910 | ||
| XM_008769677.1 | XP_008767899.1 | 6 | 2957 | |||
| XM_008769678.1 | XP_008767900.1 | 6 | 2954 | |||
| XM_008769679.1 | XP_008767901.1 | 6 | 683 | |||
| XM_008769680.1 | XP_008767902.1 | 6 | 2957 | |||
| GenBank mRNA | AF081941.1 | - | 7 | 910 | ||

Adcy10
Genomic Map