siRNA Details
| Gene Symbol: Rapgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021690.1 | NP_067722.1 | 11, 12 | 1315 | ||
| XM_006242352.2 | XP_006242414.1 | 12, 13 | 1575 | |||
| XM_008765778.1 | XP_008764000.1 | 12, 13 | 1864 | |||
| XM_008765779.1 | XP_008764001.1 | 12, 13 | 1482 | |||
| XM_008765780.1 | XP_008764002.1 | 11, 12 | 1822 | |||
| GenBank mRNA | BC107667.1 | - | 11, 12 | 1268 | 7936411 | |
| U78167.1 | - | 11, 12 | 1315 | |||

Rapgef3
Genomic Map