siRNA Details
| Gene Symbol: Sik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021693.2 | NP_067725.2 | 4 | 372 | ||
| GenBank mRNA | AB020480.1 | - | 4 | 372 | ||
| AF106937.1 | - | 430 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sik1| Gene Symbol: Sik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021693.2 | NP_067725.2 | 4 | 372 | ||
| GenBank mRNA | AB020480.1 | - | 4 | 372 | ||
| AF106937.1 | - | 430 | ||||