siRNA Details
| Gene Symbol: Arhgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021694.3 | NP_067726.3 | 27 | 2782 | ||
| GenBank mRNA | AF314539.1 | - | 25 | 2693 | ||
| AJ236911.1 | - | 25 | 2673 | |||
| BC091218.1 | - | 27 | 2782 | 7107802 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef1| Gene Symbol: Arhgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021694.3 | NP_067726.3 | 27 | 2782 | ||
| GenBank mRNA | AF314539.1 | - | 25 | 2693 | ||
| AJ236911.1 | - | 25 | 2673 | |||
| BC091218.1 | - | 27 | 2782 | 7107802 | ||