siRNA Details
| Gene Symbol: Synpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021695.2 | NP_067727.2 | 2 | 611 | ||
| GenBank mRNA | AB013130.1 | - | 1 | 137 | ||
| AM980944.1 | - | 81 | ||||
| CB794090.1 | - | 1 | 353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synpo| Gene Symbol: Synpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021695.2 | NP_067727.2 | 2 | 611 | ||
| GenBank mRNA | AB013130.1 | - | 1 | 137 | ||
| AM980944.1 | - | 81 | ||||
| CB794090.1 | - | 1 | 353 | |||