siRNA Details
| Gene Symbol: Atxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021702.1 | NP_067734.1 | 6 | 524 | ||
| XM_006240485.2 | XP_006240547.1 | 7 | 560 | |||
| XM_006240486.2 | XP_006240548.1 | 7 | 557 | |||
| XM_006240487.2 | XP_006240549.1 | 7 | 560 | |||
| XM_006240488.2 | XP_006240550.1 | 6 | 476 | |||
| XM_006240490.1 | XP_006240552.1 | 7 | 586 | |||
| XM_006240493.2 | XP_006240555.1 | 6 | 479 | |||
| GenBank mRNA | Y12319.1 | - | 6 | 524 | ||

Atxn3
Genomic Map