siRNA Details
| Gene Symbol: Akap14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021703.1 | NP_067735.1 | 6 | 1700 | ||
| GenBank mRNA | AJ002474.1 | - | 6 | 1700 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap14| Gene Symbol: Akap14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021703.1 | NP_067735.1 | 6 | 1700 | ||
| GenBank mRNA | AJ002474.1 | - | 6 | 1700 | ||