siRNA Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031658.1 | NP_113846.1 | 10, 11 | 1172 | ||
| XM_006246021.2 | XP_006246083.1 | 11, 12 | 1123 | |||
| XM_006246022.2 | XP_006246084.1 | 11, 12 | 1130 | |||
| GenBank mRNA | BC070934.1 | - | 10, 11 | 1133 | 7098161 | |
| D87351.1 | - | 10, 11 | 1172 | |||

Msln
Genomic Map