siRNA Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031658.1 | NP_113846.1 | 11 | 1315 | ||
| XM_006246021.2 | XP_006246083.1 | 12 | 1266 | |||
| XM_006246022.2 | XP_006246084.1 | 12 | 1273 | |||
| GenBank mRNA | BC070934.1 | - | 11 | 1276 | 7098161 | |
| D87351.1 | - | 11 | 1315 | |||

Msln
Genomic Map