siRNA Details
| Gene Symbol: Arpp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031660.1 | NP_113848.1 | 1, 2 | 54 | ||
| XM_006243410.2 | XP_006243472.1 | 2, 3 | 165 | |||
| GenBank mRNA | AJ005982.1 | - | 1, 2 | 54 | ||
| BC058461.1 | - | 2, 3 | 100 | 6919944 | ||
| FQ221182.1 | - | 2, 3 | 104 | |||
| FQ222843.1 | - | 2, 3 | 103 | |||
| Gene Symbol: LOC100360828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729972.4 | XP_002730018.2 | 1 | 91 | ||

Arpp19
Genomic Map