siRNA Details
| Gene Symbol: Cep78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021741.2 | NP_068509.2 | 6 | 898 | ||
| XM_006231136.2 | XP_006231198.1 | 6 | 1013 | |||
| GenBank mRNA | FQ222521.1 | - | 4 | 763 | ||
| X99330.1 | - | 6 | 686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep78| Gene Symbol: Cep78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021741.2 | NP_068509.2 | 6 | 898 | ||
| XM_006231136.2 | XP_006231198.1 | 6 | 1013 | |||
| GenBank mRNA | FQ222521.1 | - | 4 | 763 | ||
| X99330.1 | - | 6 | 686 | |||