siRNA Details
| Gene Symbol: Cep78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021741.2 | NP_068509.2 | 15 | 2289 | ||
| XM_006231136.2 | XP_006231198.1 | 15 | 2350 | |||
| GenBank mRNA | X99330.1 | - | 15 | 2023 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep78| Gene Symbol: Cep78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021741.2 | NP_068509.2 | 15 | 2289 | ||
| XM_006231136.2 | XP_006231198.1 | 15 | 2350 | |||
| GenBank mRNA | X99330.1 | - | 15 | 2023 | ||