siRNA Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021746.1 | NP_068514.1 | 11, 12 | 1211 | ||
| XM_008765781.1 | XP_008764003.1 | 11, 12 | 940 | |||
| GenBank mRNA | BC081950.1 | - | 10, 11 | 1260 | ||
| BC127549.1 | - | 3, 4 | 438 | |||
| X96488.1 | - | 11, 12 | 1211 | |||

Mapk12
Genomic Map