siRNA Details
| Gene Symbol: Acrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021747.1 | NP_068515.1 | 2 | 255 | ||
| GenBank mRNA | AJ243484.1 | - | 2 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acrv1| Gene Symbol: Acrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021747.1 | NP_068515.1 | 2 | 255 | ||
| GenBank mRNA | AJ243484.1 | - | 2 | 255 | ||