siRNA Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021748.1 | NP_068516.1 | 421 | |||
| GenBank mRNA | AF089839.1 | - | 3 | 219 | ||
| AF142097.1 | - | 421 | ||||
| AF189019.1 | - | 421 | ||||
| FQ211783.1 | - | 460 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsf| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021748.1 | NP_068516.1 | 421 | |||
| GenBank mRNA | AF089839.1 | - | 3 | 219 | ||
| AF142097.1 | - | 421 | ||||
| AF189019.1 | - | 421 | ||||
| FQ211783.1 | - | 460 | ||||