siRNA Details
| Gene Symbol: Birc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021752.2 | NP_068520.2 | 2 | 1592 | ||
| XM_006242494.2 | XP_006242556.1 | 3 | 2069 | |||
| XM_006242495.2 | XP_006242557.1 | 3 | 2270 | |||
| GenBank mRNA | AF183431.1 | - | 2 | 950 | ||
| AF190020.1 | - | 2 | 1964 | |||
| BC062055.1 | - | 2 | 1592 | 5598786 | ||

Birc2
Genomic Map