siRNA Details
| Gene Symbol: Nop58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021754.1 | NP_068522.1 | 8 | 889 | ||
| XM_006245013.1 | XP_006245075.1 | 7 | 714 | |||
| GenBank mRNA | AF194371.1 | - | 8 | 889 | ||
| BC087637.1 | - | 8 | 922 | 7309062 | ||
| FQ225580.1 | - | 1 | 85 | |||
| FQ230388.1 | - | 1 | 85 | |||

Nop58
Genomic Map