siRNA Details
| Gene Symbol: Fhit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021774.1 | NP_068542.1 | 5 | 477 | ||
| GenBank mRNA | AF170064.1 | - | 5 | 477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fhit| Gene Symbol: Fhit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021774.1 | NP_068542.1 | 5 | 477 | ||
| GenBank mRNA | AF170064.1 | - | 5 | 477 | ||