siRNA Details
| Gene Symbol: Rhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022505.2 | NP_071950.1 | 2 | 347 | ||
| XM_006239268.2 | XP_006239330.1 | 2 | 381 | |||
| GenBank mRNA | AB015191.1 | - | 2 | 312 | ||
| AF531096.1 | - | 2 | 347 | |||
| BC127471.1 | - | 2 | 336 | 8374048 | ||

Rhd
Genomic Map