siRNA Details
| Gene Symbol: Pfkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206847.1 | NP_996729.1 | 14 | 1449 | ||
| XM_008771885.1 | XP_008770107.1 | 14 | 1509 | |||
| GenBank mRNA | BC088847.1 | - | 14 | 1449 | 7312293 | |
| FN432820.1 | - | 14 | 1477 | |||
| L25387.1 | - | 14 | 1423 | |||

Pfkp
Genomic Map