siRNA Details
| Gene Symbol: Gde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032615.2 | NP_116004.2 | 472 | |||
| GenBank mRNA | AF212861.1 | - | 417 | |||
| BC070897.1 | - | 472 | 7099596 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gde1| Gene Symbol: Gde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032615.2 | NP_116004.2 | 472 | |||
| GenBank mRNA | AF212861.1 | - | 417 | |||
| BC070897.1 | - | 472 | 7099596 | |||